Learn More
Biolegend Purified anti-ZAP70 Phospho (Tyr319)/Syk Phospho (Tyr352)
Supplier:  Biolegend 683702
Encompass_Preferred
Purified anti-ZAP70 Phospho (Tyr319)/Syk Phospho (Tyr352) [1503310]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, ICFC; Size: 100 ug
Catalog No. 50-208-7474