Learn More
Biolegend Purified anti-human HLA-DR, DP, DQ
Supplier:  Biolegend 361702
Encompass_Preferred
Purified anti-human HLA-DR, DP, DQ [Tü39]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Cat (Feline), Dog (Canine), Cattle; Apps: FC, IP; Size: 100 ug
Catalog No. 50-605-385