Learn More
BINDER™ Pure Aqua Service
SDP
Catalog No. 08111127 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-127 Plastic
1 options
Catalog No. 08-111-127 Supplier BINDER™ Supplier No. 80120759
Add to Cart
Add to Cart
Pure Aqua Service system for preparation or complete desalination of tap water. For use with series KBWF, KBF, KBF P, KBF LQC, KMF, KBF-S, MKF, and MKFT.
  • Suitable for: KBF-S 720, MKF 240, KBF LQC 240, KBF P 720, KBF-S 240, KBF P 240, KBWF 720, MKFT 720, MKFT 240, MKFT 115, MKF 56, MKF 720, KBF LQC 720, KBF 720, KBWF 240, MKF 115, KMF 720, KMF 240, KMF 115, KBF 1020, KBF 240, KBF 115
Specifications
Description BINDER PURE AQUA SERVICE - System for preparation or complete desalination of tap water, complete set containing PURE AQUA 300 single-use cartridge, measuring device, and all necessary connecting parts
For Use With (Equipment) BINDER Chambers Series KBWF, KBF, KBF P, KBF LQC, KMF, KBF-S, MKF, MKFT
Material Plastic
Type Water Supply