Learn More
Biolegend Purified Armenian Hamster IgG Isotype Ctrl
Supplier:  Biolegend 400902
Encompass_Preferred
Purified Armenian Hamster IgG Isotype Ctrl [HTK888]; Isotype: Armenian Hamster IgG; Apps: FC, ICFC, IP, ICC, IHC, WB, FA; Size: 500 ug
Catalog No. NC9003345