Learn More
BUCHI Pure Essential Chromatography Fraction Rack
SDP
Catalog No. 05405939 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Seventy-five 16 x 150 mm Tubes
Seventy-five 18 x 150 mm Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-405-939 Seventy-five 18 x 150 mm Tubes
05-405-943 Seventy-five 16 x 150 mm Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-939 Supplier BUCHI Supplier No. 11074055
Add to Cart
Add to Cart
Using robust polypropylene collection racks, the fraction collector is compatible with many different sizes and designs of vessels: tubes, funnels, and flasks. With no limits for sample loading and fraction collection, this system adapts to your needs.
  • Can be equipped with two racks
  • Compatible with several different-sized glass tubes, funnels, and flasks, enabling an optimal collection depending on the fraction size
Specifications
Type Fraction Rack
For Use With (Equipment) Seventy-five 18 x 150 mm Tubes