Learn More
BUCHI Pure Essential Flash Cartridge Stand
SDP
Catalog No. 05405934 Shop All BUCHI Products
Change view
Click to view available options
Cartridge Holder
Catalog No.
05-405-934 Cartridge Holder
1 options
Catalog No. 05-405-934 Supplier BUCHI Supplier No. 11072733
Add to Cart
Add to Cart
Use this stand in combination with clamps to hold one or more cartridges ranging from 4 to 300 g.
  • Simple design
  • Supports multiple cartridge clamps
  • Stainless steel corrosion resistance
Specifications
Type Cartridge Holder
For Use With (Equipment) Chromatography System