Learn More
AVANTOR MASTERFLEX (VWR LLC) TC Tubing Clamp 3/4"" Polypropylene, 10/pk
Catalog No. NC9029739
Encompass
TC Tubing Clamp 3/4" Polypropylene, 10/pk, Install easily over existing tubing lines or assembliesSmooth contours eliminate risk of puncture or ruptureFully sterilizableMeet USP Class IV criteria
This item is not returnable. View return policy
Catalog No. NC9029739 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0682211
TC Tubing Clamp 3/4" Polypropylene, 10/pk, Install easily over existing tubing lines or assembliesSmooth contours eliminate risk of puncture or ruptureFully sterilizableMeet USP Class IV criteria