Learn More
Cytek Biosciences Inc PURE MS CD16/CD32 (2.4G2)100UG
Supplier:  Cytek Biosciences Inc 700161U100
Encompass_Preferred
Purified Anti-Mouse CD16 / CD32 (2.4G2) Isotype: Rat IgG2b Apps: FC, IF, IP Size: 100 ug
Catalog No. 50-105-5033