Learn More
Cytek Biosciences Inc PURE MS CD3E (145-2C11) 1MG
Supplier:  Cytek Biosciences Inc 700031M001
Encompass_Preferred
Purified Anti-Mouse CD3e (145-2C11) Isotype: Armenian Hamster IgG Apps: FC, IHCF, IF, IP, WB Size: 1 mg
Catalog No. 50-105-5012