Learn More
Biolegend Purified Mouse IgG2a, κ isotype Ctrl
Supplier:  Biolegend 400201
Encompass_Preferred
Purified Mouse IgG2a, κ isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: FC, ICFC, ICC, IHC, IP, WB, FA; Size: 50 ug
Catalog No. 50-168-857