Learn More
BUCHI Pure Nebulizer O-Ring
SDP
Catalog No. 05405474 Shop All BUCHI Products
Change view
Click to view available options
EPDM
Catalog No.
05-405-474 EPDM
1 options
Catalog No. 05-405-474 Supplier BUCHI Supplier No. 11066421
Add to Cart
Add to Cart
O-ring for Pure Chromatography System Nebulizers
  • For use with Pure Chromatography Systems
  • 23.52 x 1.78 mm
  • EPDM
Specifications
Material EPDM
Type O-Ring
For Use With (Equipment) Pure Chromatography System Nebulizers