Learn More
Biolegend Purified Rat IgG2b, λ Isotype Ctrl
Supplier:  Biolegend 403801
Encompass_Preferred
Purified Rat IgG2b, λ Isotype Ctrl [G013B8]; Isotype: Rat IgG2b, λ; Apps: FC; Size: 50 ug
Catalog No. 50-207-9707