Learn More
FUJIFILM Biosciences Pure Sericin
Catalog No. 5000450392
Encompass_Preferred
Sericin derived from silkworm is an effective supplement instead of FBS for mammalian cell culture medium Adding 0 1% w/v sericin to the medium promotes cell proliferation and inhibits cell death Furthermore adding 1% w/v to the basal medium allows it to be used as a serum-free freezing medium
Catalog No. 50-004-50392 Supplier FUJIFILM Biosciences Supplier No. 16722681
Sericin derived from silkworm is an effective supplement instead of FBS for mammalian cell culture medium Adding 0 1% w/v sericin to the medium promotes cell proliferation and inhibits cell death Furthermore adding 1% w/v to the basal medium allows it to be used as a serum-free freezing medium