Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Pure Solvent Bottle Platform
SDP

Catalog No. 05405540 Shop All BUCHI Products
Change view
Click to view available options
Capacity (Metric):
4 x 4 L bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
05-405-540 4 x 4 L bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-405-540 Supplier BUCHI Supplier No. 11069285
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solvent bottle platform for on top of the Pure chromatography system

  • For use with BUCHI Pure chromatography systems
  • 4 bottle (4 L) maximum

Specifications

Type Test Tube
For Use With (Application) Purification
Capacity (Metric) 4 x 4 L bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.