Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotix Pure uTIP™ Sterile Pipette Tips
GREENER_CHOICE

Catalog No. 12111161 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
250 μL
300 μL
1000 μL
1250 μL
Tip Style:
Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
12-111-161 10 μL Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
12-111-162 10 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-140 10 μL Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point, Eppendorf Style
12-111-160 200 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-159 250 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-158 300 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-157 1000 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-156 1250 μL Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 12-111-161 Supplier Biotix Supplier No. 63305413
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pure uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard are designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Manufactured with Pure resin made of virgin polypropylene.

  • Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard
  • Designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal.
  • Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin made of virgin polypropylene.
  • Fits all standard pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Hinged Rack Sterile
Tip Style Ergonomic, Extended Length, Fine Point, Finely Tapered, Flextop
Product Type Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit, Universal Fit Multichannel, Universal Fit Ultramicro
Graduated Yes
Product Line uTip
Material Polypropylene
Show More Show Less

This product contains no safety hazards

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.