Learn More
Aqua Solutions Pure Water™ pH Buffers and pHISA™ Adjustor PH 4.10, 4Pk
SDP
Catalog No. NC1768532
Encompass
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.
Catalog No. NC1768532 Supplier Aqua Solutions Supplier No. OR700402
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.