Learn More
Sigma Aldrich Fine Chemicals Biosciences Pure Water Density Standard UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 C, density: 0.9970 g/mL at 25 C | 7732-18-5 | MFCD00011332 | 60ML
Catalog No. 501809594
Encompass
Pure Water Density Standard UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 C, density: 0.9970 g/mL at 25 C | Mol Wt: 18.02 | 7732-18-5 | MFCD00011332 | 60ML
Catalog No. 50-180-9594 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. DENWAT60ML
Pure Water Density Standard UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 C, density: 0.9970 g/mL at 25 C | Mol Wt: 18.02 | 7732-18-5 | MFCD00011332 | 60ML