Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Pure xTIP Rainin LTS Compatible Racked Pipette Tips
GREENER_CHOICE

Catalog No. 12578070 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
20 μL
250 μL
300 μL
1000 μL
1200 μL
Sterility:
Non-sterile
Sterile
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
12-578-070 1200 μL Non-sterile
12-578-071 1200 μL Sterile
12-578-073 1000 μL Non-sterile
12-578-074 1000 μL Sterile
12-578-075 300 μL Non-sterile
12-578-076 300 μL Sterile
12-578-078 250 μL Non-sterile
12-578-079 250 μL Sterile
12-578-081 20 μL Non-sterile
12-578-082 20 μL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-578-070 Supplier BIOTIX™ Supplier No. 63305480
Add to Cart
Add to Cart

Designed with Flexfit™ to lower necessary insertion and ejection force by ~70% to make secure seal. Manufactured with Pure resin made of virgin polypropylene.

  • Fits Rainin LTS and x Pipette pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 3.85 in.
Length (Metric) 97.8 mm
Certifications/Compliance BSE/TSE Free, Endotoxins/Pyrogen Free, DNASE/RNASE Free, PCR Inhibitor Free
For Use With (Application) Manual Volume Transfer
Format Hinged Rack
Tip Style Ergonomic
Packaging 8 Racks/Pk, 5 Pk/Cs
Quantity 768 Tips/Pk
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated Yes
Product Line xTIP 4
Material 100% Virgin Polypropylene
Shape Standard
Graduations 200, 500, 1000
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.