Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Pure xTIP Rainin LTS Compatible Tip Eject Reload Pipette Tips
GREENER_CHOICE

Catalog No. 12578080 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
20 μL
250 μL
1000 μL
1200 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-578-080 20 μL
12-578-077 250 μL
12-578-072 1000 μL
12-578-069 1200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 12-578-080 Supplier BIOTIX™ Supplier No. 63305460
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Creates an ergonomic system in the lab with FlexFit™, StarStop™, and an LTS™ style pipette.

  • Fast manual ejection racking
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Length (English) 1.95 in.
Length (Metric) 49.5 mm
Certifications/Compliance BSE/TSE Free, Endotoxins/Pyrogen Free, DNASE/RNASE Free, PCR Inhibitor Free
For Use With (Application) Manual Volume Transfer
Sterilization Method E-Beam
Format Refill
Tip Style Ergonomic
Packaging 10 Refills/Pk, 5 Pk/Cs
Quantity 960 Tips/Pk.
Product Type Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated Yes
Product Line xTIP 4
Material 100% Virgin Polypropylene
Shape Standard
Graduations 2, 10, 20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.