Learn More
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces.
- These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Bottom Shape | Flat |
| Color | Clear |
| Culture Area | 1.9 cm² |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Cell Culture Plate |
| Well Count | 24 |
| No. per Case | 5 Plates |
| Storage Requirements | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.