Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PureCoat™ Amine Rectangular Straight Neck Cell Culture Flask with Vented Cap

Catalog No. 01100299 Shop All Corning Products
Change view
Click to view available options
Volume (Metric) Working:
175 cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Working
01-100-299 175 cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-100-299 Supplier Corning™ Supplier No. 354728
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays.

Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces.

  • These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
TRUSTED_SUSTAINABILITY

Specifications

Material Polystyrene
Neck Style Straight
No. per Case 5 Flasks
No. per Pack 5 Flasks
Shape Rectangular
Surface Coating Amine
Graduated No
Mouth Style Threaded
Sterility Sterile
Bottom Shape Flat
Closure Type Vented Cap
Culture Area 175 cm²
Volume (Metric) Working 175 cm²
Quantity 5/Cs., 5/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.