Learn More
Advanced Biomatrix PureCol, Bovine Collagen, 3 mg/mL, 100 mL, Atelocollagen
Supplier:  Advanced Biomatrix 5005100ML
Encompass_Preferred
PureCol is a 3 mg/ml, Type I bovine atelocollagen solution for 2D and 3D cell culture, or as a collagen standard. PureCol is known as the standard among collagen products for purity (≥99.9% collagen content), consistency and reproducibility, and is cited in >2000 publications. PureCol collagen is approximately 97% Type I atelocollagen with the remainder being comprised of Type III collagen. PureCol collagen is supplied at approximately a 3 mg/ml concentration. The concentration for each specific lot is provided on a Certificate of Analysis that is available with the purchase of each product. PureCol is soluble atelocollagen in 0.01 N HCI, therefore, the pH is 2. PureCol collagen is ideal for coating of surfaces, providing preparation of thin layers for culturing cells, or use as a solid gel. PureCol collagen is provided in a user-friendly packaging for use and storage. PureCol is sterile filtered and is supplied as a ready to use solution. Collagen Video: https://youtu.be/YcK2vUpNt0g
Catalog No. 50-360-230