Learn More
Sigma Aldrich Fine Chemicals Biosciences PureColTM EZ Gel solution
Catalog No. 502900881
Encompass
PureCol(TM) is used in cell culture tissue engineering and as a coating material for medical devices.
Catalog No. 50-290-0881 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 507435ML
PureCol(TM) is used in cell culture tissue engineering and as a coating material for medical devices.