Learn More
Sigma Aldrich Fine Chemicals Biosciences PureColTM S Collagen Stand
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CC300
Encompass
Collagen is the main structural protein of the extracellular matrix found within various connective tissues of the body. Type I collagen is a major structural component of skin bone tendon and other fibrous connective tissues. In order to provide the most in vivo-like environment for cells researchers have historically cultured cells inside collagen hydrogels or on top of collagen coated plasticware. PureCol(R) is an ultrapure bovine skin derived collagen that is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography providing product consistency reproducibility and is cited in over 2000 publications. PureCol(R) Coated T25 flasks have a uniform and consistent application of high quality Type I collagen on clear polystyrene surface. The vented flasks have an anti-drip edge and modified canted neck to provide maximum control replacing nutrient depleted media. The flask
Catalog No. 50-290-4975