Learn More
CUBE BIOTECH PureCube Carboxy Activated Agarose; 10 ml
Catalog No. NC2684651
Encompass
10 ml of Carboxy Activated Agarose for the coupling of biomolecules with an exposed NH2-group
Catalog No. NC2684651 Supplier CUBE BIOTECH Supplier No. 50103
10 ml of Carboxy Activated Agarose for the coupling of biomolecules with an exposed NH2-group