Learn More
CUBE BIOTECH PureCube Co-NTA MagBeads; 1 ml
Catalog No. NC2684634
Encompass
1ml of Co-NTA MagBead slurry (25%) for Protein Purification of His-tagged proteins
Catalog No. NC2684634 Supplier CUBE BIOTECH Supplier No. 31501
1ml of Co-NTA MagBead slurry (25%) for Protein Purification of His-tagged proteins