Learn More
CUBE BIOTECH PureCube Epoxy Activated Agarose; 50 ml
Catalog No. NC2684653
Encompass
50 ml of Epoxy Activated Agaroses for the coupling of biomolecules with an exposed NH2 or SH-group
Catalog No. NC2684653 Supplier CUBE BIOTECH Supplier No. 50705
50 ml of Epoxy Activated Agaroses for the coupling of biomolecules with an exposed NH2 or SH-group