Learn More
CUBE BIOTECH PureCube Glutathione MagBeads; 25 ml
Catalog No. NC2684636
Encompass
25 ml of Glutathione MagBead slurry (25%) for Protein Purification of GST-tagged proteins
Catalog No. NC2684636 Supplier CUBE BIOTECH Supplier No. 32225
25 ml of Glutathione MagBead slurry (25%) for Protein Purification of GST-tagged proteins