Learn More
CUBE BIOTECH PureCube Amine Activated Agarose; 10 ml
Catalog No. NC2582054
Encompass
10 ml of Amine Activated Agarose for the coupling of biomolecules with an exposed COOH-group
Catalog No. NC2582054 Supplier CUBE BIOTECH Supplier No. 51003
10 ml of Amine Activated Agarose for the coupling of biomolecules with an exposed COOH-group