Learn More
CUBE BIOTECH PureCube Maleimide Activated MagBeads; 5 ml
Catalog No. NC2083017
Encompass
PureCube Maleimide Activated-5ml Type:Magnetic BeadsBead size:30 ¿m (MagBeads)Purpose: Biomolecule CouplingActivated Chemical Group: Maleimide
Catalog No. NC2083017 Supplier CUBE BIOTECH Supplier No. 51205
PureCube Maleimide Activated-5ml Type:Magnetic BeadsBead size:30 ¿m (MagBeads)Purpose: Biomolecule CouplingActivated Chemical Group: Maleimide