Learn More
System Biosciences LLC PUREFECTION TRANSFECTION REAGENTPureFection Transfection Reagent = 1 mL
SDP
Supplier:  System Biosciences LLC LV750A1
Encompass_Preferred
Deliver more nucleic acids—plasmids, siRNAs, etc.—to cells than the leading lipid-based transfection reagent for effective and reproducible Lentivirus transfection. With SBI’s PureFection™ Transfection Reagent, you can deliver more nucleic acid—plasmids, siRNAs, etc.—than the leading lipid-based transfection reagent for effective, efficient, and reproducible transfections.The easy-to-use protocol consists of a rapid, one-step, 15-minute incubation with the plasmid, small RNA, or other nucleic acid you’d like to transfect. Once the incubation is done, simply add directly to target cells—no media changes are required as PureFection works in the presence of antibiotics and serum.
Catalog No. NC0377483