Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Air Inlet Adaptor
SDP

Catalog No. 18444023
Change view
Click to view available options
Model:
PF3000-03-066 BP
PF3000-03-066 MP
Quantity:
10/Pk.
250/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Quantity
18-444-023 PF3000-03-066 BP 10/Pk.
18-444-024 PF3000-03-066 MP 250/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-444-023 Supplier Gentex™ Supplier No. PF300003066BP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Air inlet adaptors keeps air-purifying protection systems running in peak condition. For use with the PureFlo 3000 PAPR.

  • For use with PureFlo 3000 PAPR
  • Color: White

Specifications

Description Air Inlet Adaptor (10-Pk.)
Color White
Disposable Single-use
Material Plastic
Product Type Air Inlet Adapter
For Use With (Equipment) PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-03-066 BP
Quantity 10/Pk.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.