Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Cleaning Plugs
SDP

Catalog No. 18444008
Change view
Click to view available options
Cleaning Plugs; Color: Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-008 Cleaning Plugs; Color: Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-008 Supplier Gentex™ Supplier No. PF300003014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Set of 3 cleaning plugs to protect the PureFlo 3000 PAPR system.

  • PureFlo 3000 Cleaning Plugs
  • For Plugging Spaces on the PF300 Unit during cleaning
  • Color: Red

Specifications

Description PureFlo 3000 Cleaning Plugs
Color Red
Disposable Reusable
Material Rubber
Product Type Cleaning and Storage Plug
For Use With (Application) Plugging Spaces on The PF300 Unit
For Use With (Equipment) PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-03-014
Quantity 3/Pk.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.