Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Ear Defenders
SDP

Catalog No. 18444027
Change view
Click to view available options
Ear Defenders; Color: Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-027 Ear Defenders; Color: Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-027 Supplier Gentex™ Supplier No. PF300004001

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ear Defenders (NRR 22) for use with the PureFlo 3000 PAPR.

  • Hearing protection with NRR of 22
  • For use with PureFlo 3000 PAPR

Specifications

Description Ear Defenders NRR 22
Color Red
Disposable Reusable
Product Type Ear Defenders
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-04-001
Quantity 1 Ea.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.