Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Faceshield
SDP

Catalog No. 18444014
Change view
Click to view available options
Faceshield; Color: Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-014 Faceshield; Color: Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-014 Supplier Gentex™ Supplier No. PF300003044
Add to Cart
Add to Cart
This item is not returnable. View return policy

Faceshield with optional tear-off layers for use with the PureFlo 3000 PAPR.

  • Face Protection
  • For use with PureFlo 3000 PAPR

Specifications

Description Faceshield
Color Clear
Disposable Reusable
Product Type Visor
Size One Size
Facepiece Type Faceshield
For Use With (Equipment) PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-03-044
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.