Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Hard Hat
SDP

Catalog No. 18444025
Change view
Click to view available options
Hard Hat; Color: Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-025 Hard Hat; Color: Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-025 Supplier Gentex™ Supplier No. PF300003067
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blue Hard Hat for use with the PureFlo 3000 PAPR.

  • Hard Hat
  • For use with PureFlo 3000 PAPR
  • Blue

Specifications

Description Hard Hat - Blue
Color Blue
Material Plastic
Product Type Hardhat Assembly
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-03-067
Quantity 1 Ea.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.