Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Headband
SDP

Catalog No. 18444019
Change view
Click to view available options
Headband; For use with: PureFlo 3000 PAPR; Color: Black; Includes: 3 Comfort Pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-019 Headband; For use with: PureFlo 3000 PAPR; Color: Black; Includes: 3 Comfort Pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-019 Supplier Gentex™ Supplier No. PF300003058
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Headband for use with the PureFlo 3000 PAPR.

  • Headband for use with PureFlo 3000 PAPR
  • Includes: 3 Comfort Pads

Specifications

Description Headband with clips and straps
Color Black
Disposable Reusable
Material Plastic
Product Type Headband
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Includes 3 comfort pads
Model PF3000-03-057
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.