Learn More
Gentex™ PureFlo 3000 PAPR (NIOSH Approved) with Overhood
SDP
Catalog No. 18444034
Change view
Click to view available options
Model:
PF3000-F6NA-03
Includes:
Open Frame, Tychem QC Hood
Catalog No. Model Includes
18-444-034 PF3000-F6NA-03 Open Frame, Tychem QC Hood
1 options
Catalog No. 18-444-034 Supplier Gentex™ Supplier No. PF3000F6NA03
Add to Cart
Add to Cart
Innovative all-in-one respiratory and face protection with enhanced comfort and performance.
  • The PureFlo 3000 is a modular, lightweight PAPR that provides superior protection in light-to-medium duty industrial environments.
  • An all-in-one respirator optimized for maximum mobility, the PureFlo 3000 integrates respiratory, eye, and face protection into one easy-to use unit.
  • The head-mounted design eliminates the need for hoses and belts, virtually eliminating snag hazards in tight quarters.
  • A breakthrough in headborne weight, comfort, and durability, the PureFlo 3000 is made for keeping workforces safe, while simultaneously enhancing productivity.
  • Used in a wide variety of applications including healthcare, pharmaceutical manufacturing and clean room environments.
Specifications
ANSI Standard HH: ANSI/ISEA Z89.1:2014 Type 1 Class G ; FACESHIELD - ANSI/ISEA Z87.1:2015 Impact Rated (Z87+)
Assigned Protection Factor (APF) 1000
Battery Life 4 to 5 hours
Battery Type Lithium-ion
Cartridge/Filter PARTICULATE
Certifications (NIOSH) NIOSH Approved
Product Type PAPR System with Hood
Size One Size
Facepiece/Headtop Model PF3000-03-044
Filter Efficiency High Efficiency Particulate Air (HEPA) Filter
Filter Efficiency (NIOSH) HEPA
Includes Open Frame, Tychem QC Hood
Model PF3000-F6NA-03
Noise Level Less than 80 dBA
Quantity 1 Ea.
Recharge Time 2 hr.
Replaceable Filter Yes
With Faceseal Yes