Learn More
Gentex™ PureFlo 3000 PAPR
SDP
Catalog No. 18444035
Change view
Click to view available options
Model:
PF3000-H2SB-03
Includes:
Blue Hard Hat, Clear Visor, Standard Face Seal
Catalog No. Model Includes
18-444-035 PF3000-H2SB-03 Blue Hard Hat, Clear Visor, Standard Face Seal
1 options
This item is not returnable. View return policy
Catalog No. 18-444-035 Supplier Gentex™ Supplier No. PF3000H2SB03
Add to Cart
Add to Cart
This item is not returnable. View return policy
NIOSH-approved air-purifying protection systems offer all-in-one respiratory, eye, face, and head protection. Enhanced features for maximum mobility and simplified use make it ideal in pharmaceutical manufacturing and cleanroom environments.
  • Modular, lightweight PAPR that provides protection in light-to-medium duty industrial environments
  • All-in-one protection integrates respiratory, eye, and face protection into one easy-to use unit
  • Head-mounted design eliminates the need for hoses and belts, eliminating snag hazards in tight quarters
  • Used in a wide variety of applications including healthcare, pharmaceutical manufacturing and cleanroom environments
Specifications
ANSI Standard HH: ANSI/ISEA Z89.1:2014 Type 1 Class G ; FACESHIELD - ANSI/ISEA Z87.1:2015 Impact Rated (Z87+)
Assigned Protection Factor (APF) 25
Battery Life 4 to 5 hours
Battery Type Lithium-ion
Cartridge/Filter PARTICULATE
Certifications (NIOSH) NIOSH Approved
Product Type PAPR Hard Hat System
Size One Size
Facepiece/Headtop Model PF3000-03-044
Filter Efficiency High Efficiency Particulate Air (HEPA) Filter
Filter Efficiency (NIOSH) HEPA
Includes Blue Hard Hat, Clear Visor, Standard Face Seal
Model PF3000-H2SB-03
Noise Level Less than 80 dBA
Quantity 1 Ea.
Recharge Time 2 hr.
Replaceable Filter Yes
With Faceseal Yes