Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 PAPR Storage Bag
SDP

Catalog No. 18444002
Change view
Click to view available options
Bag; Color: Black; Size: One-Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-002 Bag; Color: Black; Size: One-Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-002 Supplier Gentex™ Supplier No. PF300003046
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Storage/carry bag for the PureFlo 3000 PAPR.

  • Helmet Bag for use with PureFlo ESM+ and PureFlo 3000 PAPR
  • Made of Durable Tear-Resistant Nylon Shell

Specifications

Description GID_900000014704362
Color Black
Disposable Reusable
Material Durable Tear-resistant Nylon Shell
Product Type Carry Bag
Size One Size
For Use With (Equipment) PureFlo ESM+ and PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-03-046
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.