Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Tychem QC Disposable Slipover Hood
SDP

Catalog No. 18444012
Change view
Click to view available options
Hood; Color: White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-012 Hood; Color: White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-012 Supplier Gentex™ Supplier No. PF300003023
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pack of 10 disposable Tychem QC slipover hoods for use with the PureFlo 3000 PAPR.

  • Tychem QC Disposable Slip-Over Hood
  • Extra protection for PureFlo 3000 PAPR

Specifications

Color White
Material Tychem
Product Type Slipover Hood
Size One Size
For Use With (Application) Extra protection for PureFlo 3000 PAPR
For Use With (Equipment) PureFlo 3000 PAPR
Model PF3000-03-023
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.