Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Tychem QC Faceseal
SDP

Catalog No. 18444013
Change view
Click to view available options
Face Seal; Material: Tychem; Color: Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-013 Face Seal; Material: Tychem; Color: Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-013 Supplier Gentex™ Supplier No. PF300003024
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pack of 10 Tychem QC faceseals for use with the PureFlo 3000 PAPR.

  • Face Seal for PureFlo 3000 PAPR
  • Yellow
  • Tychem Material

Specifications

Description Tychem QC faceseal (10-Pk.)
Color Yellow
Disposable Reusable
Material Tychem
Product Type Faceseal
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Model PF3000-03-024
Quantity 10/Pk.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.