Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Visor Assembly
SDP

Catalog No. 18444015
Change view
Click to view available options
Visor Assembly; For Use With: PureFlo 3000 PAPR; Color: Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-015 Visor Assembly; For Use With: PureFlo 3000 PAPR; Color: Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-015 Supplier Gentex™ Supplier No. PF300003070
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Visor assembly including faceshield, frame, clips and mounting hardware for the PureFlo 3000 PAPR.

  • Visor for PureFlo 3000 PAPR
  • Includes: Faceshield, Frame, Clips and Mounting Hardware

Specifications

Description Visor Assembly
Color Clear
Disposable Reusable
Product Type Visor
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Includes Faceshield, Frame, Clips and Mounting Hardware
Latex Free Yes
Model PF3000-03-070
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.