Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 Visor Protectors
SDP

Catalog No. 18444031
Change view
Click to view available options
Visor Protectors; For Use With: PureFlo 3000 PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-031 Visor Protectors; For Use With: PureFlo 3000 PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-031 Supplier Gentex™ Supplier No. PF300004022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Visor protectors (4-pack) for use with the PureFlo 3000 PAPR.

  • Accessory for PureFlo 3000 PAPR

Specifications

Description Visor protector
Color Clear
Disposable Single-use
Product Type Visor Protector
Size One Size
For Use With (Equipment) PureFlo 3000 PAPR
Latex Free Yes
Model PF3000-04-022
Quantity 4/Pk.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.