Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Blower Assembly
SDP

Catalog No. 17010061
Change view
Click to view available options
Flow Rate:
8.1 CFM
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
17-010-061 8.1 CFM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-010-061 Supplier Gentex™ Supplier No. PR05723SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spare Blower Assembly for the PureFlo ESM+ PAPR

  • Unique air circulation system creates a smooth continuous flow of air from blower through filter located at rear of helmet
  • The advanced electronics package within the ESM+ continually monitors all functions and provides a self-diagnostic system check to ensure proper functioning.
  • The improved electronics also provide an increased flow rate at 8.1 CFM/230Lpm.
  • Contains a fully capsuled blower module with a single 7-way IP67* connector between the sidebars and blower/motor unit
  • Standard internal on/off switch, in addition to having an external power switch option like the standard ESM

Specifications

Color Gray
Product Type Motor Blower
Flow Rate 8.1 CFM
Model PR05723SP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.