Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ HE/HF/HC Filter
SDP

Catalog No. 18444045
Change view
Click to view available options
Filter; Color: Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-045 Filter; Color: Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-045 Supplier Gentex™ Supplier No. PR02026SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

ESM+ Filter HE/HF/HC Filter for use with PureFlo ESM+ PAPR.

  • ESM+ Filter HE/HF/HC Filter
  • For use with PureFlo ESM+ PAPR
  • NIOSH Approved

Specifications

Cartridge/Filter PARTICULATE
Certifications (NIOSH) NIOSH Approved
Color Gray
Filter Type HE/HF/HC
Product Type Filter
Filter Efficiency High Efficiency Particulate Air (HEPA), Hydrogen Fluoride (HF), and Hydrogen Chloride (HC), Filter Cartridge provides nuisance level removal against most Organic Vapors, Inorganic vapors, Acid Gas and O-zone.
For Use With (Equipment) PureFlo ESM+ PAPR
Latex Free Yes
Quantity 1 Ea.
Replaceable Filter Yes
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.