Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Headband Upgrade Kit
SDP

Catalog No. 18444041
Change view
Click to view available options
Headband Kit; Color: Gray; For Use With: PureFlo ESM+ PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-041 Headband Kit; Color: Gray; For Use With: PureFlo ESM+ PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-041 Supplier Gentex™ Supplier No. PR01602SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PureFlo ESM+ quick release headband upgrade kit includes new headband PR01600SP, brow pad and nape pad, left and right mounting kit.

  • Upgrade Kit

Specifications

Description Quick Release Headband Upgrade Kit
Color Gray
Disposable Reusable
Material Plastic
Product Type Headband
Size One Size
For Use With (Equipment) PureFlo ESM+ PAPR
Includes Headband, brow and nape pad, left and right mounting kit
Latex Free Yes
Model PR01602SP
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.