Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Headband
SDP

Catalog No. 18444040
Change view
Click to view available options
Headband; Material: Plastic; Color: Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-040 Headband; Material: Plastic; Color: Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-040 Supplier Gentex™ Supplier No. PR01600SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Headband assembly for use with the PureFlo ESM+, includes 3 comfort pads.

  • Accessory for PureFlo ESM+ PAPR

Specifications

Description ESM Headband Assembly
Color Gray
Disposable Reusable
Material Plastic
Product Type Headband
Size One Size
For Use With (Equipment) PureFlo ESM+ PAPR
Includes 3 Comfort Pads
Latex Free Yes
Model PR01600SP
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.