Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Interface Cable
SDP

Catalog No. 18444058
Change view
Click to view available options
Interface Cable; For Use With: PureFlo ESM+ PAPR; Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-058 Interface Cable; For Use With: PureFlo ESM+ PAPR; Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-058 Supplier Gentex™ Supplier No. PR05708SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Interface bracket assembly for PureFlo ESM+ PAPR

  • Accessory for PureFlo ESM+ PAPR

Specifications

Description Interface Bracket Assembly
Color Black
Disposable Reusable
Material Plastic
Product Type Interface Cable
Size One Size
For Use With (Equipment) PureFlo ESM+ PAPR
Latex Free Yes
Model PR05708SP
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.