Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo PRO2475 Fire Resistant Neck Cape
SDP

Catalog No. 18444052
Change view
Click to view available options
Model:
PR02475
PR02475SP
Quantity:
1 Ea.
2/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Quantity
18-444-052 PR02475 1 Ea.
18-444-053 PR02475SP 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-444-052 Supplier Gentex™ Supplier No. PR02475
Add to Cart
Add to Cart
This item is not returnable. View return policy

Spare neck cape assembly for use with the PureFlo ESM+ PAPR.

  • Flame Resistant as per ASTM D6413
  • Abrasion Resistant
  • Flash Fire Protection as per ASTM F1930
  • Chemical Penetration Resistance as per ASTM F903
  • Adjustable neck cord
  • Two velcro attachments for securing free end of lanyard
  • Twelve snap fasteners for easy removal and attachment
  • Used with PureFlo ESM, ESM+ and Airline SAR

Specifications

Color Black
Disposable Reusable
Material Stedprene
Product Type Neck Cape
Size One Size
For Use With (Equipment) PureFlo ESM+, ESM, and PR02000 PAPR Helmet Systems
Includes Adjustable neck cord
Model PR02475
Quantity 1 Ea.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.